Zinc phosphide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trizinc diphosphide, Zinc phosphide
045459-02
045459-04
045459-09
AA45459-02EA
101
USD
AA45459-02
AA45459-04
AA45459-09
Zinc phosphide ≥99.999% (metals basis)
Zinc phosphide
Purity: 99.999%
CAS No.: 1314-84-7
Molecular Formula: Zn3P2
Formula Weight: 258.10
Physical Form: 6mm & down pieces
Density: 4.55
Melting Point: 420°
Boiling Point: 1100°
Merck Reference: 14,10152
UN No.: 1714
MDL No.: MFCD00049631
Formula:
P₂Zn₃ MW: 258.12 Boiling Pt: 1100 Melting Pt: 420 Density: 4.55 Storage Temperature: Ambient |
MDL Number:
MFCD00049631 CAS Number: 1314-84-7 EINECS: 215-244-5 UN: 1714 ADR: 4.3 I Merck Index: 14,10152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...