Silver cyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Argentous cyanate

045411-06 045411-14
AA45411-06EA 77.2 USD
AA45411-06 AA45411-14 AA45411-22
Silver cyanate 98%
Silver cyanate

Purity: 98% CAS No.: 3315-16-0 Molecular Formula: CAgNO Formula Weight: 149.89 Physical Form: Powder Density: 4.0 Appearance: White to grey Odor: Odorless Beilstein Reference: 3655040 UN No.: 3077 MDL No.: MFCD00011512

Formula: CAgNO
MW: 149.89 g/mol
Density: 4.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011512
CAS Number: 3315-16-0
EINECS: 222-006-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR