Gallium(III) chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gallium trichloride
045026-06
045026-14
045026-22
AA45026-06EA
78.4
USD
AA45026-06
AA45026-14
AA45026-22
Gallium(III) chloride, ultra dry ≥99.999% (metals basis)
Gallium(III) chloride
Purity: 99.999%
CAS No.: 13450-90-3
Molecular Formula: GaCl3
Formula Weight: 176.03
Physical Form: 8mm and down Granules, Ampouled under argon
Density: 2.47
Appearance: White
Melting Point: 78°
Boiling Point: 201°
Merck Reference: 14,4348
UN No.: 3260
MDL No.: MFCD00011018
Formula:
Cl₃Ga MW: 176.08 g/mol Boiling Pt: 225 Melting Pt: 77.9 Density: 2.47 Storage Temperature: Ambient |
MDL Number:
MFCD00011018 CAS Number: 13450-90-3 EINECS: 236-610-0 UN: 3260 ADR: 8 II Merck Index: 14,4348 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...