Titanium carbide, nanopowder

Supplier: Thermo Scientific Chemicals
Warning

044922-06 044922-14 044922-22
AA44922-06EA 51.9 USD
AA44922-06 AA44922-14 AA44922-22
Titanium carbide, nanopowder
Titanium carbide

CAS No.: 12070-08-5 Molecular Formula: CTi Formula Weight: 59.91 Physical Form: 30-50nm APS Powder, S.A. 35-45m2/g Density: 4.93 Appearance: Black Melting Point: ca 3140° Boiling Point: 4820° UN No.: 3178 MDL No.: MFCD00011268

Formula: CTi
MW: 59.89 g/mol
Boiling Pt: 4820 °C
Melting Pt: ∼ 3140 °C
Density: 4.93
Storage Temperature: Ambient
MDL Number: MFCD00011268
CAS Number: 12070-08-5
EINECS: 235-120-4
UN: 3178
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR