Lead selenide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead(II) selenide
044916-14
044916-22
AA44916-14EA
241
USD
AA44916-14
AA44916-22
Lead selenide ≥99.999% (metals basis)
Lead selenide
Purity: 99.999+%
CAS No.: 12069-00-0
Molecular Formula: PbSe
Formula Weight: 286.15
Physical Form: 3-6mm Fused Lump
Odor: Product specific
UN No.: 3077
MDL No.: MFCD00016273
Formula:
PbSe MW: 286.16 g/mol Melting Pt: 79.5…80 °C Density: 8.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016273 CAS Number: 12069-00-0 EINECS: 235-109-4 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...