Zinc peroxide ∼ 50% ZnO₂

Supplier: Thermo Scientific Chemicals
Danger

039635-22 039635-36
AA39635-22EA 37.85 USD
AA39635-22 AA39635-36
Zinc peroxide ∼ 50% ZnO₂
Zinc peroxide

Applications: Curing agent, rubber accelerator


CAS No.: 1314-22-3 Molecular Formula: ZnO2 Formula Weight: 97.38 Physical Form: Powder, remainder ZnO Odor: Odorless Melting Point: 150° dec. UN No.: 1516 MDL No.: MFCD00137469


Curing agent, rubber accelerator

Formula: O₂Zn
MW: 97.39
Melting Pt: 212 (dec.)
Density: 1.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00137469
CAS Number: 1314-22-3
EINECS: 215-226-7
UN: 1516
ADR: 5.1 II
Merck Index: 14,10149

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR