Alizarine yellow R (sodium salt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
038707-09
038707-14
AA38707-09EA
70.8
USD
AA38707-09
AA38707-14
Alizarine yellow R (sodium salt)
Alizarine Yellow R (sodium salt)
Transition interval: pH 10.1 (yellow) to pH 12.1 (orange)
CAS No.: 1718-34-9
Molecular Formula: C13H8N3NaO5
Formula Weight: 309.22
Physical Form: Powder
Appearance: Dark yellow to brown
Odor: Odorless
Beilstein Reference: 962365
Merck Reference: 14,255
MDL No.: MFCD00067121
Formula:
C₁₃H₈N₃NaO₅ MW: 309.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067121 CAS Number: 1718-34-9 EINECS: 217-002-4 Merck Index: 14,255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...