Zirconium(IV) ethoxide 99%

Supplier: Thermo Scientific Chemicals
Warning

018859-18 018859-09
AA18859-18EA 418 USD
AA18859-18 AA18859-09
Zirconium(IV) ethoxide 99%
Zirconium(IV) ethoxide

Purity: 99% CAS No.: 18267-08-8 Molecular Formula: Zr(OC2H5)4 Formula Weight: 271.47 Physical Form: Crystalline Sensitivity: Moisture Sensitive Boiling Point: 235°/5mm UN No.: 3181 MDL No.: MFCD00015654

Formula: C₈H₂₀O₄Zr
MW: 271.47 g/mol
Boiling Pt: 235 °C (5 mmHg)
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00015654
CAS Number: 18267-08-8
UN: 3181
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR