Gold trihydroxide Au ≥ 79% (≥79% Au)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Gold(iii)hydroxide
012159-03
012159-06
012159-09
AA12159-03EA
226
USD
AA12159-03
AA12159-06
AA12159-09
Gold trihydroxide Au ≥ 79% (≥79% Au)
Gold trihydroxide
Applications: Gold plating solutions, porcelain decoration
Decomposed by light to metallic gold
CAS No.: 1303-52-2
Molecular Formula: Au(OH)3
Formula Weight: 247.99
Physical Form: Powder
Sensitivity: Light Sensitive
Odor: Odorless
Merck Reference: 14,4523
MDL No.: MFCD00046173
Gold plating solutions, porcelain decoration
Formula:
H₃AuO₃ MW: 247.99 Storage Temperature: Ambient |
MDL Number:
MFCD00046173 CAS Number: 1303-52-2 EINECS: 215-120-0 Merck Index: 14,4523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...