Titanium diboride

Supplier: Thermo Scientific Chemicals

011364-30 011364-A1 011364-18
AA11364-30EA 167 USD
AA11364-30 AA11364-A1 AA11364-18
Titanium diboride
Titanium diboride

Applications: aircraft industry, in cutting and grinding applications, as protective coatings for metals in high temperature applications, as a base material for cermet production
Soluble in hydrogen peroxide and sulfuric acid


CAS No.: 12045-63-5 Molecular Formula: B2Ti Formula Weight: 69.50 Physical Form: -325 Mesh Powder Density: 4.38 Odor: Odorless MDL No.: MFCD00049595


In the aircraft industry, in cutting and grinding applications, as protective coatings for metals in high temperature applications, as a base material for cermet production

Formula: B₂Ti
MW: 69.5 g/mol
Melting Pt: 3225 °C
Density: 4,38
Storage Temperature: Ambient
MDL Number: MFCD00049595
CAS Number: 12045-63-5
EINECS: 234-961-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR