Ammonium molybdate (para) hydrate ≥99.999% (metals basis excluding W), Puratronic® (max. 300 ppm W)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium molybdate hydrate

010811-06 010811-14
AA10811-06EA 60.1 USD
AA10811-06 AA10811-14
Ammonium molybdate (para) hydrate ≥99.999% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Ammonium molybdate (para) hydrate

Fieser: 21,19


Purity: 99.999% CAS No.: 12027-67-7 Molecular Formula: H24Mo7N6O24.xH2O Formula Weight: 1163.79(anhy) Physical Form: Powder Density: 2.498 Appearance: White Melting Point: ca 190° dec. Merck Reference: 14,533 MDL No.: MFCD00064633

Formula: H₂₄Mo₇N₆O₂₄
MW: 1163.8 g/mol (anhy)
Melting Pt: ∼ 190 °C (dec.)
Density: 2.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064633
CAS Number: 12027-67-7
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR