Anti-POSTN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-E09 600-401-E10
RL600-401-E09EA 656.12 USD
RL600-401-E09 RL600-401-E10
Anti-POSTN Rabbit Polyclonal Antibody
Antibodies
Anti-Periostin Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a three bands of approximately 93 kDa in size corresponding to periostin pan Proteins in mouse lung extract.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100;

Type: Primary
Antigen: POSTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR