Anti-DBH Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
600-601-D27 600-601-D28
RL600-601-D27EA 656.12 USD
RL600-601-D27 RL600-601-D28
Anti-DBH Sheep Polyclonal Antibody
Antibodies
Anti-Dopamine beta-Hydroxylase Antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optmized by the end user. Expect a band of approximately 75 kDa in size corresponding to DBH Protein.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Sheep
Isotype: IgG
Reactivity: Human, non-Human Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR