Condenser C and CR Assemblies for Rotary Evaporators, Ace Glass Incorporated
Supplier: Ace Glass
3937-04 3938-01 3938-100 3938-03 3937-06 3937-05 3938-10 3937-112 3937-01 3937-12 3937-03 3937-100 3937-10 3955-08 3955-03
89083-038EA 420.52 USD
89083-038 89083-040 89083-042 89083-068 89083-072 89083-074 89083-086 89083-112 89083-114 89083-116 89083-118 89083-120 89083-122 89083-124 89083-126
Condenser C and CR Assemblies for Rotary Evaporators, Ace Glass Incorporated
Condensers
Both assemblies fit models 200/205 and series 114-144 rotary evaporators.
  • Both sets are available poly coated, or standard non-coated
  • Both sets are fabricated from borosilicate glass
The “C” Assembly contains the main glass outer trap body with a 35/20 inner ball joint bottom connection that attaches to the receiver flasks (which can be purchased separately). Features standard top and side flange that connects directly to the rotary evaporator. Assembly “C” also includes the glass cold trap inner body, part #00672, PTFE sealing ring with Viton o-rings, PTFE lid or cap, PTFE hose barbs, and GL threaded caps.
The “CR” Assembly is very similar to the “C” Assembly, except that the bottom is a [ST] 45/50 inner joint which fits into a separate distribution head. The complete “CR” Assembly includes the main glass cold trap outer body, glass cold trap inner body, PTFE cap, PTFE seal ring with Viton o-rings, GL thread caps, and hose barbs.
Información para pedidos: Complete assemblies, or components may be purchased separately. Both assemblies include 89083-150 and 89083-152 stopcocks, 89083-098 joint adapter (without clips), and 89083-096 glass steam tube. Stainless steel pinch clamp (89060-258) for receiving flask is also included in 89083-116 and 89083-112.