Anti-Tyrosine Hydroxylase Sheep Polyclonal Antibody

Supplier: Novus Biologicals
NB300-110 NB300-110SS
102129-920EA 589.54 USD
102129-920 102129-922
Anti-Tyrosine Hydroxylase Sheep Polyclonal Antibody
Antibodies
The Tyrosine Hydroxylase Antibody from Novus Biologicals is a sheep polyclonal antibody to Tyrosine Hydroxylase. This antibody reacts with human, mouse, rat, avian, mammal, rabbit, reptile. The Tyrosine Hydroxylase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat, Avian, Mammal, Rabbit, Reptile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR