Anti-Gastric Lipase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-60930 NB110-60930SS
102124-256EA 503.38 USD
102124-256 102124-270
Anti-Gastric Lipase Rabbit Polyclonal Antibody
Antibodies
The Gastric Lipase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Gastric Lipase. This antibody reacts with human, mouse. The Gastric Lipase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Gastric Lipase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR