Anti-S-arrestin Mouse Monoclonal Antibody [clone: PDS-1]

Supplier: Novus Biologicals
NB100-2385 NB100-2385SS
102107-308EA 503.38 USD
102107-308 102107-320
Anti-S-arrestin Mouse Monoclonal Antibody [clone: PDS-1]
Antibodies
The S-arrestin Antibody (PDS-1) from Novus Biologicals is a mouse monoclonal antibody to S-arrestin. This antibody reacts with human, porcine, bovine. The S-arrestin Antibody (PDS-1) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: S-arrestin
Clonality: Monoclonal
Clone: PDS-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR