Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody [clone: SE-1]

Supplier: Novus Biologicals
NB110-68095 NB110-68095SS
102124-618EA 491.08 USD
102124-618 102124-620
Anti-Hepatic Sinusoidal Endothelial Cells Mouse Monoclonal Antibody [clone: SE-1]
Antibodies
Mouse Monoclonal Hepatic Sinusoidal Endothelial Cells Antibody (SE-1). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1-5 ?g/ml

Type: Primary
Antigen: Hepatic Sinusoidal Endothelial Cells
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR