Anti-ABCA1 Mouse Monoclonal Antibody [clone: HJ1]

Supplier: Novus Biologicals
NB100-2068 NB100-2068SS
102106-586EA 528 USD
102106-586 102106-598
Anti-ABCA1 Mouse Monoclonal Antibody [clone: HJ1]
Antibodies
The ABCA1 Antibody (HJ1) from Novus Biologicals is a mouse monoclonal antibody to ABCA1. This antibody reacts with human, mouse, rat, rabbit. The ABCA1 Antibody (HJ1) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

CyTOF-ready

Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: HJ1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR