Lymphoma Markers Antibody Pack

Supplier: Novus Biologicals
NBP1-78950
102213-530EA 306.15 USD
102213-530
Lymphoma Markers Antibody Pack
Antibodies
Lymphoma Markers Antibody Pack. Tested Applications: Western Blot. Tested Reactivity: Human, Rat.

This Lymphoma Markers Antibodies Sample Pack includes antibodies to EPB41L3 and CADM1.

Type:
Antigen: EPB41L3+CADM1
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR