Anti-IgM Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-72748
102203-150EA 281.66 USD
102203-150
Anti-IgM Goat Polyclonal Antibody
Antibodies
Goat Polyclonal Goat anti-Ferret IgM Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Ferret.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR