Geldanamycin compound ≥95%

Supplier: Novus Biologicals

Synonyms: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

NB110-81559
102125-506EA 352.43 USD
102125-506
Geldanamycin compound ≥95%
Geldanamycin

Geldanamycin is a benzoquinoid ansamycin produced by Streptomyces hygroscopicus. It binds specifically to heat shock protein HSP90 and downregulates target proteins including tyrosine kinases, steroid receptors, transcription factors and cell cycle regulatory kinases (1,2). It induces the inactivation, destabilization and eventual degradation of HIF-1


  • Slightly soluble in methanol, chloroform or DMSO (10mg/mL)
  • Insoluble in water
  • Yellow solid


Precaución: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Formula: C₂₉H₄₀N₂O₉
MW: 560.64 g/mol
MDL Number: MFCD00274570
CAS Number: 30562-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR