Anti-SR-AI/MSR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-00093SS NBP1-00093
102154-540EA 208 USD
102154-540 102154-526
Anti-SR-AI/MSR Rabbit Polyclonal Antibody
Antibodies
The SR-AI / MSR Antibody from Novus Biologicals is a rabbit polyclonal antibody to SR-AI / MSR. This antibody reacts with human, mouse. The SR-AI / MSR Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SR-AI/MSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR