Anti-BRCA1 Mouse Monoclonal Antibody [clone: RAY]

Supplier: Novus Biologicals
NB100-598 NB100-598SS
102110-792EA 503.38 USD
102110-792 102110-862
Anti-BRCA1 Mouse Monoclonal Antibody [clone: RAY]
Antibodies
The BRCA1 Antibody (RAY) from Novus Biologicals is a mouse monoclonal antibody to BRCA1. This antibody reacts with human. The BRCA1 Antibody (RAY) has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, CyTOF-ready.

Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: RAY
Conjugation: Unconjugated
Epitope: The epitope recognized is found between amino acids 1314-1600.
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR