1-Fluoro-4-ethynylbenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne

069647-25G 069647-1G
101780-862EA 273.85 USD
101780-862 101780-858
1-Fluoro-4-ethynylbenzene ≥98%
1-Fluoro-4-ethynylbenzene

Hazard: Flammable Solid/Irritant/Refrigerate

Formula: C₈H₅F
MW: 120.13
Boiling Pt: 55…56 °C
Melting Pt: 27…28 °C
Flash Pt: 29 °C
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR