2-(Trifluoromethyl)-p-anisidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-2-(trifluoromethyl)aniline

066905-5G 066905-1G
101780-848EA 50.77 USD
101780-848 101780-846
2-(Trifluoromethyl)-p-anisidine ≥98%
2-(Trifluoromethyl)-p-anisidine

Hazard: Toxic

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD04972660
CAS Number: 53903-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR