1-Butylimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

064012-100G 064012-25G
101780-824EA 67.69 USD
101780-824 101780-822
1-Butylimidazole ≥95%
1-Butylimidazole

Hazard: Toxic

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 257 °C
Density: 0.948
Flash Pt: 145 °C
MDL Number: MFCD00042753
CAS Number: 4316-42-1
UN: 2690
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR