Ethyl propiolate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester

063172-100G 063172-25G
101780-818EA 316.92 USD
101780-818 101780-816
Ethyl propiolate ≥98%
Ethyl propiolate

Hazard: Flammable/Toxic/Refrigerate/Lachrymator

Formula: C₅H₆O₂
Boiling Pt: 118…120 °C
Density: 0.963
Flash Pt: 23 °C
MDL Number: MFCD00009184
CAS Number: 623-47-2
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR