Dimethyl fluoromalonate ≥98%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-fluoromalonate

059893-25G 059893-100G
101780-812EA 110.77 USD
101780-812 101780-814
Dimethyl fluoromalonate ≥98%
Dimethyl fluoromalonate

Hazard: Toxic/Irritant-Harmful

Formula: C₅H₇FO₄
MW: 150.11 g/mol
Boiling Pt: 111…112 °C (45 mmHg)
MDL Number: MFCD00054680
CAS Number: 344-14-9

Order Now

Specification Test Results

Boiling point 111-112°/4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR