2-Chloro-4-(trifluoromethoxy)aniline ≥97%

Supplier: Matrix Scientific

059784-25G 059784-5G
101780-808EA 136.92 USD
101780-808 101780-806
2-Chloro-4-(trifluoromethoxy)aniline ≥97%
2-Chloro-4-(trifluoromethoxy)aniline

Hazard: Toxic/Irritant

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD01631704
CAS Number: 69695-61-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR