2-Chloropyridine-3-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

058903-1G 058903-5G
101780-790EA 60 USD
101780-790 101780-792
2-Chloropyridine-3-sulfonyl chloride ≥95%
2-Chloropyridine-3-sulfonyl chloride

Hazard: Corrosive/Irritant

Formula: C₅H₃Cl₂NO₂S
Melting Pt: 42…43 °C
MDL Number: MFCD05664916
CAS Number: 6684-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR