2,2-Difluoro-1,3-benzodioxole ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole

058597-5G 058597-25G
101780-782EA 16.92 USD
101780-782 101780-784
2,2-Difluoro-1,3-benzodioxole ≥97%
2,2-Difluoro-1,3-benzodioxole

Hazard: Flammable/Irritant

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 129…130 °C
Density: 1.308
Flash Pt: 32 °C
MDL Number: MFCD00236217
CAS Number: 1583-59-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 129-130°
Density 1.308

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR