4-Nitro-1,2-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

058508-250MG 058508-1G
101780-774EA 85.54 USD
101780-774 101780-776
4-Nitro-1,2-bis(trifluoromethyl)benzene ≥95%
4-Nitro-1,2-bis(trifluoromethyl)benzene

Hazard: Irritant-Harmful

Formula: C₈H₃F₆NO₂
Boiling Pt: 78 °C (6 Torr)
MDL Number: MFCD00728841
CAS Number: 1978-20-7

Order Now

Specification Test Results

Boiling point 78°C@6torr


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR