4-Ethoxy-1,1,1-trifluorobut-3-en-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Ethoxyvinyl) trifluoromethyl ketone, (3E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 1-Ethoxy-3-oxo-4,4,4-trifluorobut-1-ene

057760-5G 057760-25G
101780-768EA 61.54 USD
101780-768 101780-770
4-Ethoxy-1,1,1-trifluorobut-3-en-2-one ≥95%
4-Ethoxy-1,1,1-trifluorobut-3-en-2-one

Hazard: Flammable/Refrigerate

Formula: C₆H₇F₃O₂
MW: 168.115 g/mol
Boiling Pt: 51…53 °C (12 mmHg)
Flash Pt: 52 °C
MDL Number: MFCD00192131
CAS Number: 17129-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR