4-Methoxybenzylamine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

057114-100G 057114-500G
101780-754EA 75.38 USD
101780-754 102833-200
4-Methoxybenzylamine ≥98%
4-Methoxybenzylamine

Hazard: Corrosive/Irritant/Air Sensitive

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 236…237 °C
Density: 1.05
MDL Number: MFCD00008122
CAS Number: 2393-23-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Boiling point 236-237°
Density 1.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR