2,4-Dichlorobenzylamine
Supplier: Matrix Scientific
057003-50G
057003-10G
101780-746EA
424.62
USD
101780-746
101780-744
2,4-Dichlorobenzylamine
2,4-Dichlorobenzylamine
Hazard: Corrosive
Formula:
C₇H₇Cl₂N Boiling Pt: 83…84 °C (5 mmHg) Density: 1.32 |
MDL Number:
MFCD00008109 CAS Number: 95-00-1 UN: 2735 ADR: 8,III |
Specification Test Results
Boiling point | 83-84°/5mm |
Density | 1.32 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...