2,4-Dichlorobenzonitrile ≥98%
Supplier: Matrix Scientific
057001-25G
057001-100G
057001-5G
101780-736EA
247.69
USD
101780-736
101780-738
101780-734
2,4-Dichlorobenzonitrile ≥98%
2,4-Dichlorobenzonitrile
Hazard: Toxic
Formula:
C₇H₃Cl₂N Melting Pt: 61°C |
MDL Number:
MFCD00016373 CAS Number: 6574-98-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Melting point | 61° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...