2,3-Dichlorobenzonitrile ≥98%
Supplier: Matrix Scientific
057000-5G
057000-25G
101780-730EA
190.77
USD
101780-730
101780-732
2,3-Dichlorobenzonitrile ≥98%
2,3-Dichlorobenzonitrile
Hazard: Toxic
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Boiling Pt: 114°C/6mm Melting Pt: 61-63°C |
MDL Number:
MFCD00016372 CAS Number: 6574-97-6 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 114°/6mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...