Cyclohexanemethyl isothiocyanate ≥99%

Supplier: Matrix Scientific

056989-5G 056989-25G
101780-722EA 203.08 USD
101780-722 101780-724
Cyclohexanemethyl isothiocyanate ≥99%
Cyclohexanemethyl isothiocyanate

Hazard: Toxic/Corrosive

Formula: C₈H₁₃NS
Boiling Pt: 222…223 °C
Density: 1.02
MDL Number: MFCD00040881
CAS Number: 52395-66-1

Order Now

Specification Test Results

Boiling point 222-223°
Density 1.02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR