4-Butylbenzylamine

Supplier: Matrix Scientific

Synonyms: (4-Butylphenyl)methanamine

056938-5G 056938-25G 056938-100G
101780-692EA 338.46 USD
101780-692 101780-694 101780-696
4-Butylbenzylamine
4-Butylbenzylamine

Hazard: Corrosive

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 134 °C (10 mmHg)
Density: 0.93
MDL Number: MFCD02258855
CAS Number: 57802-79-6

Order Now

Specification Test Results

Boiling point 134°/10mm
Density 0.93

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR