2-Aminopyridine ≥99%

Supplier: Matrix Scientific

056889-250G 056889-1KG
101780-682EA 58.46 USD
101780-682 101780-684
2-Aminopyridine ≥99%
2-Aminopyridine

Hazard: Toxic

Formula: C₅H₆N₂
Melting Pt: 58…60 °C
MDL Number: MFCD00006312
CAS Number: 504-29-0
UN: 2671
ADR: 6.1,II

Order Now

Specification Test Results

Melting point 58-60°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR