Allyl isothiocyanate ≥98%
Supplier: Matrix Scientific
056881-25G
056881-100G
101780-674EA
36.92
USD
101780-674
101780-676
Allyl isothiocyanate ≥98%
Allyl isothiocyanate
Hazard: Flammable/Toxic/Marine Pollutant/Moisture Sensitive/Refrigerate
Formula:
C₄H₅NS MW: 99.16 g/mol Boiling Pt: 150°C Density: 1.01 |
MDL Number:
MFCD00004822 CAS Number: 57-06-7 UN: 1545 ADR: 6.1,II |
Specification Test Results
Boiling point | 150° |
Density | 1.01 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...