1-Bromo-4-(1,1-difluoroethyl)benzene

Supplier: Matrix Scientific

054391-1G 054391-5G
101780-654EA 110.77 USD
101780-654 101780-656
1-Bromo-4-(1,1-difluoroethyl)benzene
1-Bromo-4-(1,1-difluoroethyl)benzene

Hazard: Toxic/Irritant

Formula: C₈H₇BrF₂
MDL Number: MFCD11110333
CAS Number: 1000994-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR