4-Cyano-3,5-dichloropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloro-4-pyridinecarbonitrile, 3,5-Dichloropyridine-4-carbonitrile, 3,5-Dichloroisonicotinonitrile

048729-5G 048729-25G
101780-642EA 109.23 USD
101780-642 101780-644
4-Cyano-3,5-dichloropyridine ≥98%
4-Cyano-3,5-dichloropyridine

Hazard: Toxic/Irritant

Formula: C₆H₂Cl₂N₂
Melting Pt: 115…119 °C
MDL Number: MFCD00104651
CAS Number: 153463-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR