4-Bromo-1-butene ≥98%
Supplier: Matrix Scientific
048493-25G
048493-100G
101780-638EA
30.77
USD
101780-638
101780-640
4-Bromo-1-butene ≥98%
4-Bromo-1-butene
Hazard: Flammable/Lachrymator
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 98…100 °C Density: 1.33 |
MDL Number:
MFCD00000258 CAS Number: 5162-44-7 UN: 1992 ADR: 3,II |
Specification Test Results
Boiling point | 98-100° |
Density | 1.33 |
ND20 | 1.4625 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...