Phenethyl isocyanate ≥95%
Supplier: Matrix Scientific
038681-5G
038681-25G
101780-608EA
44.62
USD
101780-608
101780-610
Phenethyl isocyanate ≥95%
Phenethyl isocyanate
Hazard: Toxic
Formula:
C₉H₉NO Boiling Pt: 210 °C Density: 1.063 |
MDL Number:
MFCD00037044 CAS Number: 1943-82-4 |
Specification Test Results
Boiling point | 210°C |
Density | 1.063 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...