Benzyl isocyanate ≥98%
Supplier: Matrix Scientific
038672-5G
038672-25G
101780-604EA
58.46
USD
101780-604
101780-606
Benzyl isocyanate ≥98%
Benzyl isocyanate
Hazard: Flammable/Toxic/Moisture Sensitive/Refrigerate/Lachrymator
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 101…104 °C (33 mmHg) Density: 1.07 Flash Pt: 45 °C |
MDL Number:
MFCD00009701 CAS Number: 3173-56-6 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...