Thiophene-2-ethylamine ≥95%
Supplier: Matrix Scientific
037593-100G
037593-25G
101780-592EA
106.15
USD
101780-592
101780-590
Thiophene-2-ethylamine ≥95%
Thiophene-2-ethylamine
Hazard: Corrosive/Air Sensitive
Formula:
C₆H₉NS Boiling Pt: 201°C Density: 1.087 Flash Pt: 87°C |
MDL Number:
MFCD00051495 CAS Number: 30433-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...