1-Propanesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: Propane-1-sulphonyl chloride

037470-25G 037470-100G
101780-586EA 27.69 USD
101780-586 101780-588
1-Propanesulfonyl chloride ≥98%
1-Propanesulphonyl chloride

Hazard: Corrosive/Irritant

Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 78-9°C/15mm
Density: 1.267
Flash Pt: 91°C
MDL Number: MFCD00007462
CAS Number: 10147-36-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 78-9°/15mm
Density 1.267
ND20 1.453

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR