Cyclopropanemethylamine ≥95%

Supplier: Matrix Scientific

037104-25G 037104-5G
101780-580EA 192.31 USD
101780-580 101780-578
Cyclopropanemethylamine ≥95%
Cyclopropanemethylamine

Hazard: Flammable/Irritant

Formula: C₄H₉N
Boiling Pt: 84…85 °C
Density: 0.82
MDL Number: MFCD00037147
CAS Number: 2516-47-4
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Boiling point 84-85 ºC
Density 0.82


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR